missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. C24841G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24841G 1 g
C24845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24841G Supplier TCI America Supplier No. C24841G

Chemical Identifiers

CAS 206362-80-3
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.47
MDL Number MFCD04115859
InChI Key CNUYBEIHDWMLSD-UHFFFAOYSA-N
Synonym 4-bromomethyl-1-chloro-2-fluorobenzene, 4-chloro-3-fluorobenzyl bromide, 3-fluoro-4-chlorobenzyl bromide, 3-fluoro-4-chlorobenzylbromide, 4-chloro-3-fluorobenzylbromide, alpha-bromo-4-chloro-3-fluorotoluene, 4-chloro-3 fluoro benzyl bromide, benzene, 4-bromomethyl-1-chloro-2-fluoro, pubchem4898, acmc-209fcn
PubChem CID 2783136
IUPAC Name 4-(bromomethyl)-1-chloro-2-fluorobenzene
SMILES FC1=C(Cl)C=CC(CBr)=C1

Specifications

Melting Point 33°C
Quantity 1 g
UN Number 3261
Formula Weight 223.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Chloro-3-fluorobenzyl Bromide