accessibility menu, dialog, popup

UserName

BrFCl

CAS RN 206362-80-3

IUPAC Name: 4-(bromomethyl)-1-chloro-2-fluorobenzene
Synonyms: 4-(bromomethyl)-1-chloro-2-fluorobenzene
Molecular Weight (g/mol): 223.47
Molecular Formula: C7H5BrClF
InChi Key: CNUYBEIHDWMLSD-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=CC(CBr)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)