missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzyl bromide, 97%

Catalog No. AAH2639103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2639103 1 g
AAH2639106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2639103 Supplier Thermo Scientific Chemicals Supplier No. H2639103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 206362-80-3 | C7H5BrClF | 223.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 206362-80-3
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.47
MDL Number MFCD04115859
InChI Key CNUYBEIHDWMLSD-UHFFFAOYSA-N
Synonym 4-bromomethyl-1-chloro-2-fluorobenzene, 4-chloro-3-fluorobenzyl bromide, 3-fluoro-4-chlorobenzyl bromide, 3-fluoro-4-chlorobenzylbromide, 4-chloro-3-fluorobenzylbromide, alpha-bromo-4-chloro-3-fluorotoluene, 4-chloro-3 fluoro benzyl bromide, benzene, 4-bromomethyl-1-chloro-2-fluoro, pubchem4898, acmc-209fcn
PubChem CID 2783136
IUPAC Name 4-(bromomethyl)-1-chloro-2-fluorobenzene
SMILES FC1=C(Cl)C=CC(CBr)=C1

Specifications

Melting Point 30°C to 33°C
Quantity 1 g
UN Number UN3261
Formula Weight 223.48
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-fluorobenzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only