missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7125895
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32161G 1 g
C32165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32161G Supplier TCI America Supplier No. C32161G
Only null left

Chemical Identifiers

CAS 32137-20-5
Molecular Formula C7H3ClF4
Molecular Weight (g/mol) 198.54
MDL Number MFCD00060646
InChI Key HKBWIKAFHCGRTH-UHFFFAOYSA-N
Synonym 4-chloro-3-fluorobenzotrifluoride, 1-chloro-2-fluoro-4-trifluoromethyl benzene, 3-fluoro-4-chlorobenzotrifluoride, 4-chloro-3-fluoroobenzotrifluoride, benzene, 1-chloro-2-fluoro-4-trifluoromethyl, 1-chloro-2-fluoro-4-trifluoromethyl-benzene, 4-chloro-3-fluoro benzotrifluoride, 3-fluoro-4-chloro-trifluoromethyl benzene, 1-chloranyl-2-fluoranyl-4-trifluoromethyl benzene
PubChem CID 2736545
IUPAC Name 1-chloro-2-fluoro-4-(trifluoromethyl)benzene
SMILES FC1=C(Cl)C=CC(=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 135°C
Quantity 1 g
UN Number 1993
Formula Weight 198.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chloro-3-fluorobenzotrifluoride
EINECSNumber (3)-4052
TSCA No