CAS RN 32137-20-5
IUPAC Name:
1-chloro-2-fluoro-4-(trifluoromethyl)benzene
Synonyms:
1-chloro-2-fluoro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol):
198.54
Molecular Formula:
C7H3ClF4
InChi Key:
HKBWIKAFHCGRTH-UHFFFAOYSA-N
SMILES:
FC1=C(Cl)C=CC(=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Chloro-3-fluorobenzotrifluoride 98.0+%, TCI America™
CAS: 32137-20-5 Molecular Formula: C7H3ClF4 Molecular Weight (g/mol): 198.54 MDL Number: MFCD00060646 InChI Key: HKBWIKAFHCGRTH-UHFFFAOYSA-N Synonym: 4-chloro-3-fluorobenzotrifluoride,1-chloro-2-fluoro-4-trifluoromethyl benzene,3-fluoro-4-chlorobenzotrifluoride,4-chloro-3-fluoroobenzotrifluoride,benzene, 1-chloro-2-fluoro-4-trifluoromethyl,1-chloro-2-fluoro-4-trifluoromethyl-benzene,4-chloro-3-fluoro benzotrifluoride,3-fluoro-4-chloro-trifluoromethyl benzene,1-chloranyl-2-fluoranyl-4-trifluoromethyl benzene PubChem CID: 2736545 IUPAC Name: 1-chloro-2-fluoro-4-(trifluoromethyl)benzene SMILES: FC1=C(Cl)C=CC(=C1)C(F)(F)F