missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3,5-dinitrobenzotrifluoride 98.0+%, TCI America™
$128.62 - $240.46
Chemical Identifiers
| CAS | 393-75-9 |
|---|---|
| Molecular Formula | C7H2ClF3N2O4 |
| Molecular Weight (g/mol) | 270.548 |
| MDL Number | MFCD00007068 |
| InChI Key | HFHAVERNVFNSHL-UHFFFAOYSA-N |
| Synonym | 4-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,3-dinitro-5-trifluoromethyl benzene, 3,5-dinitro-4-chlorobenzotrifluoride, benzene, 2-chloro-1,3-dinitro-5-trifluoromethyl, 1-chloro-2,6-dinitro-4-trifluoromethyl benzene, 1,3-dinitro-2-chloro-5-trifluoromethylbenzene, cdnt, chloralin, 2,6-dinitro-4-trifluoromethylchlorobenzene, trifluoromethyl-3,5-dinitro-4-chlorobenzene |
| PubChem CID | 9809 |
| IUPAC Name | 2-chloro-1,3-dinitro-5-(trifluoromethyl)benzene |
| SMILES | C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(F)(F)F |
Chemical Identifiers
| 393-75-9 | |
| 270.548 | |
| HFHAVERNVFNSHL-UHFFFAOYSA-N | |
| 9809 | |
| C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(F)(F)F |
| C7H2ClF3N2O4 | |
| MFCD00007068 | |
| 4-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,3-dinitro-5-trifluoromethyl benzene, 3,5-dinitro-4-chlorobenzotrifluoride, benzene, 2-chloro-1,3-dinitro-5-trifluoromethyl, 1-chloro-2,6-dinitro-4-trifluoromethyl benzene, 1,3-dinitro-2-chloro-5-trifluoromethylbenzene, cdnt, chloralin, 2,6-dinitro-4-trifluoromethylchlorobenzene, trifluoromethyl-3,5-dinitro-4-chlorobenzene | |
| 2-chloro-1,3-dinitro-5-(trifluoromethyl)benzene |
Specifications
| 393-75-9 | |
| Yellow | |
| MFCD00007068 | |
| 2811 | |
| HFHAVERNVFNSHL-UHFFFAOYSA-N | |
| 2-chloro-1,3-dinitro-5-(trifluoromethyl)benzene | |
| 9809 | |
| ≥98.0% (GC) | |
| 4-Chloro-3,5-dinitrobenzotrifluoride |
| 56°C | |
| C7H2ClF3N2O4 | |
| 10 g | |
| 4-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,3-dinitro-5-trifluoromethyl benzene, 3,5-dinitro-4-chlorobenzotrifluoride, benzene, 2-chloro-1,3-dinitro-5-trifluoromethyl, 1-chloro-2,6-dinitro-4-trifluoromethyl benzene, 1,3-dinitro-2-chloro-5-trifluoromethylbenzene, cdnt, chloralin, 2,6-dinitro-4-trifluoromethylchlorobenzene, trifluoromethyl-3,5-dinitro-4-chlorobenzene | |
| C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(F)(F)F | |
| 270.548 | |
| 270.55 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XS9065000
TSCA : Yes