missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3,5-dinitrobenzotrifluoride 98.0+%, TCI America™
Supplier: TCI America C099425G
Specifications
| 4-Chloro-3,5-dinitrobenzotrifluoride | |
| 56°C | |
| C7H2ClF3N2O4 | |
| 25 g | |
| 4-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,3-dinitro-5-trifluoromethyl benzene, 3,5-dinitro-4-chlorobenzotrifluoride, benzene, 2-chloro-1,3-dinitro-5-trifluoromethyl, 1-chloro-2,6-dinitro-4-trifluoromethyl benzene, 1,3-dinitro-2-chloro-5-trifluoromethylbenzene, cdnt, chloralin, 2,6-dinitro-4-trifluoromethylchlorobenzene, trifluoromethyl-3,5-dinitro-4-chlorobenzene | |
| C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(F)(F)F | |
| 270.548 | |
| 270.55 | |
| Crystalline Powder |
| 393-75-9 | |
| Yellow | |
| MFCD00007068 | |
| 2811 | |
| HFHAVERNVFNSHL-UHFFFAOYSA-N | |
| 2-chloro-1,3-dinitro-5-(trifluoromethyl)benzene | |
| 9809 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 393-75-9 | |
| 270.548 | |
| HFHAVERNVFNSHL-UHFFFAOYSA-N | |
| 9809 | |
| C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(F)(F)F |
| C7H2ClF3N2O4 | |
| MFCD00007068 | |
| 4-chloro-3,5-dinitrobenzotrifluoride, 2-chloro-1,3-dinitro-5-trifluoromethyl benzene, 3,5-dinitro-4-chlorobenzotrifluoride, benzene, 2-chloro-1,3-dinitro-5-trifluoromethyl, 1-chloro-2,6-dinitro-4-trifluoromethyl benzene, 1,3-dinitro-2-chloro-5-trifluoromethylbenzene, cdnt, chloralin, 2,6-dinitro-4-trifluoromethylchlorobenzene, trifluoromethyl-3,5-dinitro-4-chlorobenzene | |
| 2-chloro-1,3-dinitro-5-(trifluoromethyl)benzene |
Safety and Handling
RTECSNumber : XS9065000
TSCA : Yes