missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dinitrobenzoic acid, 97%

Catalog No. p-7082098
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1237314 25 g
AAA1237322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1237314 Supplier Thermo Scientific Chemicals Supplier No. A1237314
Only null left

CAS: 118-97-8 | C7H3ClN2O6 | 246.559 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 118-97-8
Molecular Formula C7H3ClN2O6
Molecular Weight (g/mol) 246.559
MDL Number MFCD00007080
InChI Key PCTFIHOVQYYAMH-UHFFFAOYSA-N
Synonym 4-chloro-3,5-dinitrobenzoic acid, 3,5-dinitro-4-chlorobenzoic acid, benzoic acid, 4-chloro-3,5-dinitro, 4-chloro-3,5-dinitrobenzoate, 4-chloro-3,5-dinitro-benzoic acid, benzoicacid, 4-chloro-3,5-dinitro, pubchem22980, acmc-1bt9j, ksc180k5t, timtec-bb sbb003176
PubChem CID 8377
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])Cl)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 159°C to 162°C
Boiling Point 315°C
Flash Point 186°C (367°F)
Quantity 25 g
Beilstein 668253
Formula Weight 246.56
Percent Purity 99%
Chemical Name or Material 4-Chloro-3,5-dinitrobenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 204-290-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only