missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™

Catalog No. C32855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32855G 5 g
C32851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C32855G Supplier TCI America Supplier No. C32855G

Chemical Identifiers

CAS 142994-09-0
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.563
MDL Number MFCD03094162
InChI Key RKZXXMAQKMOZLK-UHFFFAOYSA-N
Synonym 4-chloro-2-trifluoromethyl benzoic acid, benzoic acid, 4-chloro-2-trifluoromethyl, intermediates-zcf02089, acmc-20a63a, 4-chloro-2-trifluoromethyl benzoicacid, 4-chloro-alpha,alpha,alpha-trifluoro-o-toluic acid
PubChem CID 2773856
IUPAC Name 4-chloro-2-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)C(=O)O

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 5 g
Formula Weight 224.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-(trifluoromethyl)benzoic Acid