CAS RN 142994-09-0
IUPAC Name:
4-chloro-2-(trifluoromethyl)benzoic acid
Synonyms:
4-chloro-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
224.56
Molecular Formula:
C8H4ClF3O2
InChi Key:
RKZXXMAQKMOZLK-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(Cl)C=C1C(F)(F)F
1
–
1
of
1
results
1
4-Chloro-2-(trifluoromethyl)benzoic Acid 98.0+%, TCI America™
CAS: 142994-09-0 Molecular Formula: C8H4ClF3O2 Molecular Weight (g/mol): 224.563 MDL Number: MFCD03094162 InChI Key: RKZXXMAQKMOZLK-UHFFFAOYSA-N Synonym: 4-chloro-2-trifluoromethyl benzoic acid,benzoic acid, 4-chloro-2-trifluoromethyl,intermediates-zcf02089,acmc-20a63a,4-chloro-2-trifluoromethyl benzoicacid,4-chloro-alpha,alpha,alpha-trifluoro-o-toluic acid PubChem CID: 2773856 IUPAC Name: 4-chloro-2-(trifluoromethyl)benzoic acid SMILES: C1=CC(=C(C=C1Cl)C(F)(F)F)C(=O)O