missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methylphenol, 97%

Catalog No. AAL0416406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0416406 5 g
AAL0416414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0416406 Supplier Thermo Scientific Chemicals Supplier No. L0416406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1570-64-5 | C7H7ClO | 142.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1570-64-5
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00002321
InChI Key RHPUJHQBPORFGV-UHFFFAOYSA-N
Synonym 4-chloro-o-cresol, 2-methyl-4-chlorophenol, 4-chloro-2-cresol, p-chloro-o-cresol, phenol, 4-chloro-2-methyl, o-cresol, 4-chloro, 4-chloro-2-methyl phenol, 4-chloro-2-methyl-phenol, 5-chloro-2-hydroxytoluene, unii-297v63w9ri
PubChem CID 14855
ChEBI CHEBI:1800
IUPAC Name 4-chloro-2-methylphenol
SMILES CC1=C(C=CC(=C1)Cl)O

Specifications

Melting Point 45°C to 48°C
Density 1.2
Boiling Point 220°C to 225°C
Flash Point 78°C (172°F)
Quantity 5 g
UN Number UN3437
Beilstein 1906684
Formula Weight 142.59
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methylphenol
Hazard Category H314-H331
Hazard Statement GHS H Statement
H331-H314
Toxic if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID
EINECSNumber 216-381-3
RTECSNumber GO7120000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only