missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog No. AAH2756214
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2756214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2756214 Supplier Thermo Scientific Chemicals Supplier No. H2756214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 55685-75-1 | Molecular Formula: C8H9ClO2 | Molecular Weight: 172.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 55685-75-1
Molecular Formula C8H9ClO2
Molecular Weight (g/mol) 172.61
MDL Number MFCD02683547
InChI Key AXXPZAITCCIOIA-UHFFFAOYSA-N
Synonym 4-chloro-2-methoxybenzyl alcohol, 4-chloro-2-methoxyphenyl methanol, 4-chloro-2-methoxy-phenyl-methanol, benzenemethanol, 4-chloro-2-methoxy, 4-chloro-2-methoxybenzyl alcohol 97
PubChem CID 3549393
IUPAC Name (4-chloro-2-methoxyphenyl)methanol
SMILES COC1=C(CO)C=CC(Cl)=C1

Specifications

Melting Point 46°C to 50°C
Flash Point >110°C (230°F)
Quantity 25 g
Formula Weight 172.61
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-methoxybenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only