accessibility menu, dialog, popup

UserName

CAS RN 55685-75-1

IUPAC Name: (4-chloro-2-methoxyphenyl)methanol
Synonyms: (4-chloro-2-methoxyphenyl)methanol
Molecular Weight (g/mol): 172.61
Molecular Formula: C8H9ClO2
InChi Key: AXXPZAITCCIOIA-UHFFFAOYSA-N
SMILES: COC1=C(CO)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results