missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-methoxybenzoic Acid 98.0+%, TCI America™
$87.60 - $260.97
Chemical Identifiers
| CAS | 57479-70-6 |
|---|---|
| Molecular Formula | C8H6ClO3 |
| Molecular Weight (g/mol) | 185.58 |
| MDL Number | MFCD00002532 |
| InChI Key | UFEYMXHWIHFRBX-UHFFFAOYSA-M |
| Synonym | 4-chloro-o-anisic acid, 2-methoxy-4-chlorobenzoic acid, 4-chloro-2-methoxy-benzoic acid, 4-chloro-2-methoxybenzoicacid, benzoic acid, 4-chloro-2-methoxy, 4-chloro-ortho-anisic acid, 2-methoxy-4-chloro benzoic acid, acmc-1axaj, 4-chloro-2-anisic acid, ksc494s8r |
| PubChem CID | 93682 |
| IUPAC Name | 4-chloro-2-methoxybenzoate |
| SMILES | COC1=CC(Cl)=CC=C1C([O-])=O |
Chemical Identifiers
| 57479-70-6 | |
| 185.58 | |
| UFEYMXHWIHFRBX-UHFFFAOYSA-M | |
| 93682 | |
| COC1=CC(Cl)=CC=C1C([O-])=O |
| C8H6ClO3 | |
| MFCD00002532 | |
| 4-chloro-o-anisic acid, 2-methoxy-4-chlorobenzoic acid, 4-chloro-2-methoxy-benzoic acid, 4-chloro-2-methoxybenzoicacid, benzoic acid, 4-chloro-2-methoxy, 4-chloro-ortho-anisic acid, 2-methoxy-4-chloro benzoic acid, acmc-1axaj, 4-chloro-2-anisic acid, ksc494s8r | |
| 4-chloro-2-methoxybenzoate |
Specifications
| 57479-70-6 | |
| White-Yellow | |
| MFCD00002532 | |
| 4-chloro-o-anisic acid, 2-methoxy-4-chlorobenzoic acid, 4-chloro-2-methoxy-benzoic acid, 4-chloro-2-methoxybenzoicacid, benzoic acid, 4-chloro-2-methoxy, 4-chloro-ortho-anisic acid, 2-methoxy-4-chloro benzoic acid, acmc-1axaj, 4-chloro-2-anisic acid, ksc494s8r | |
| COC1=CC(Cl)=CC=C1C([O-])=O | |
| 185.58 | |
| 186.59 | |
| Crystalline Powder |
| 148°C | |
| C8H6ClO3 | |
| 5 g | |
| UFEYMXHWIHFRBX-UHFFFAOYSA-M | |
| 4-chloro-2-methoxybenzoate | |
| 93682 | |
| ≥98.0% (GC,T) | |
| 4-Chloro-2-methoxybenzoic Acid |
Safety and Handling
TSCA : No