CAS RN 57479-70-6
IUPAC Name:
4-chloro-2-methoxybenzoate
Synonyms:
4-chloro-2-methoxybenzoate
Molecular Weight (g/mol):
185.58
Molecular Formula:
C8H6ClO3
InChi Key:
UFEYMXHWIHFRBX-UHFFFAOYSA-M
SMILES:
COC1=CC(Cl)=CC=C1C([O-])=O
1
–
1
of
1
results
1
4-Chloro-2-methoxybenzoic Acid 98.0+%, TCI America™
CAS: 57479-70-6 Molecular Formula: C8H6ClO3 Molecular Weight (g/mol): 185.58 MDL Number: MFCD00002532 InChI Key: UFEYMXHWIHFRBX-UHFFFAOYSA-M Synonym: 4-chloro-o-anisic acid,2-methoxy-4-chlorobenzoic acid,4-chloro-2-methoxy-benzoic acid,4-chloro-2-methoxybenzoicacid,benzoic acid, 4-chloro-2-methoxy,4-chloro-ortho-anisic acid,2-methoxy-4-chloro benzoic acid,acmc-1axaj,4-chloro-2-anisic acid,ksc494s8r PubChem CID: 93682 IUPAC Name: 4-chloro-2-methoxybenzoate SMILES: COC1=CC(Cl)=CC=C1C([O-])=O