missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. C20765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20765G 5 g
C207625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20765G Supplier TCI America Supplier No. C20765G
Only null left

Chemical Identifiers

CAS 57479-70-6
Molecular Formula C8H6ClO3
Molecular Weight (g/mol) 185.58
MDL Number MFCD00002532
InChI Key UFEYMXHWIHFRBX-UHFFFAOYSA-M
Synonym 4-chloro-o-anisic acid, 2-methoxy-4-chlorobenzoic acid, 4-chloro-2-methoxy-benzoic acid, 4-chloro-2-methoxybenzoicacid, benzoic acid, 4-chloro-2-methoxy, 4-chloro-ortho-anisic acid, 2-methoxy-4-chloro benzoic acid, acmc-1axaj, 4-chloro-2-anisic acid, ksc494s8r
PubChem CID 93682
IUPAC Name 4-chloro-2-methoxybenzoate
SMILES COC1=CC(Cl)=CC=C1C([O-])=O

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 5 g
Formula Weight 186.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-methoxybenzoic Acid