missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-iodoanisole 98.0+%, TCI America™

Catalog No. C22985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22985G 5 g
C229825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22985G Supplier TCI America Supplier No. C22985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 52807-27-9
Molecular Formula C7H6ClIO
Molecular Weight (g/mol) 268.48
MDL Number MFCD00052858
InChI Key CXXXAGIYCFOGHA-UHFFFAOYSA-N
Synonym 4-chloro-2-iodoanisole, 5-chloro-2-methoxyiodobenzene, 4-chloro-2-iodo-1-methoxy-benzene, 4-chloroiodoanisole, pubchem14310, 2-iodo-4-chloroanisole, acmc-209l1x, 4-chloro-2-iodo-1-methoxybenzen, benzene,4-chloro-2-iodo-1-methoxy, 4-chloro-2-iodoanisol;chloro-2-iodoanisole
PubChem CID 2800978
IUPAC Name 4-chloro-2-iodo-1-methoxybenzene
SMILES COC1=CC=C(Cl)C=C1I

Specifications

Melting Point 47°C
Color White-Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 268.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-iodoanisole