accessibility menu, dialog, popup

UserName

CAS RN 52807-27-9

IUPAC Name: 4-chloro-2-iodo-1-methoxybenzene
Synonyms: 4-chloro-2-iodo-1-methoxybenzene
Molecular Weight (g/mol): 268.48
Molecular Formula: C7H6ClIO
InChi Key: CXXXAGIYCFOGHA-UHFFFAOYSA-N
SMILES: COC1=CC=C(Cl)C=C1I
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results