missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-iodoaniline 98.0+%, TCI America™

Catalog No. C18005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18005G 5 g
C180025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18005G Supplier TCI America Supplier No. C18005G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63069-48-7
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.467
MDL Number MFCD01863737
InChI Key FLEJOBRWKBPUOX-UHFFFAOYSA-N
Synonym 2-iodo-4-chloroaniline, 4-chloro-2-iodo-aniline, 4-chloro-2-iodo-phenylamine, 4-chloro-2-iodophenylamine, benzenamine, 4-chloro-2-iodo, zlchem 532, pubchem3645, 4-chloro-6-iodoaniline, acmc-1bcyk, 2-iodo-4-chloro-aniline
PubChem CID 2757615
IUPAC Name 4-chloro-2-iodoaniline
SMILES C1=CC(=C(C=C1Cl)I)N

Specifications

Melting Point 41°C
Quantity 5 g
UN Number 2811
Formula Weight 253.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2-iodoaniline