accessibility menu, dialog, popup

UserName

CAS RN 63069-48-7

IUPAC Name: 4-chloro-2-iodoaniline
Synonyms: 4-chloro-2-iodoaniline
Molecular Weight (g/mol): 253.47
Molecular Formula: C6H5ClIN
InChi Key: FLEJOBRWKBPUOX-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1I
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)