missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzylamine hydrochloride, 97%

Catalog No. AAL1348806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1348806 5 g
AAL1348803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1348806 Supplier Thermo Scientific Chemicals Supplier No. L1348806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 202982-63-6 | C7H8Cl2FN | 196.046 g/mol

4-Chloro-2-fluorobenzylamine hydrochloride It is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-2-fluorobenzylamine hydrochloride It is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 202982-63-6
Molecular Formula C7H8Cl2FN
Molecular Weight (g/mol) 196.046
MDL Number MFCD00143286
InChI Key DFRJZBWKVAXYRV-UHFFFAOYSA-N
Synonym 4-chloro-2-fluorobenzylamine hydrochloride, 4-chloro-2-fluorophenyl methanamine hydrochloride, 4-chloro-2-fluorobenzylamine hcl, 4-chloro-2-fluorophenyl methylamine, chloride, 1-4-chloro-2-fluorophenyl methanamine hydrochloride, pubchem3428, acmc-1cp68, 4-chloro-2-fluoro-benzylamine hydrochloride
PubChem CID 2724909
IUPAC Name (4-chloro-2-fluorophenyl)methanamine;hydrochloride
SMILES C1=CC(=C(C=C1Cl)F)CN.Cl

Specifications

Melting Point 243°C to 245°C
Quantity 5 g
Solubility Information It is soluble in water.
Formula Weight 196.05
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-fluorobenzylamine hydrochloride
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only