missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-fluorobenzaldehyde 98.0+%, TCI America™
$149.13 - $443.65
Chemical Identifiers
| CAS | 61072-56-8 |
|---|---|
| Molecular Formula | C7H4ClFO |
| Molecular Weight (g/mol) | 158.556 |
| MDL Number | MFCD00143282 |
| InChI Key | UVGYSEIWAOOIJR-UHFFFAOYSA-N |
| Synonym | 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985 |
| PubChem CID | 2724908 |
| IUPAC Name | 4-chloro-2-fluorobenzaldehyde |
| SMILES | C1=CC(=C(C=C1Cl)F)C=O |
Chemical Identifiers
| 61072-56-8 | |
| 158.556 | |
| UVGYSEIWAOOIJR-UHFFFAOYSA-N | |
| 2724908 | |
| C1=CC(=C(C=C1Cl)F)C=O |
| C7H4ClFO | |
| MFCD00143282 | |
| 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985 | |
| 4-chloro-2-fluorobenzaldehyde |
Specifications
| 61072-56-8 | |
| White-Yellow | |
| MFCD00143282 | |
| 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985 | |
| C1=CC(=C(C=C1Cl)F)C=O | |
| 158.556 | |
| 158.56 | |
| Crystalline Powder |
| 60°C | |
| C7H4ClFO | |
| 5 g | |
| UVGYSEIWAOOIJR-UHFFFAOYSA-N | |
| 4-chloro-2-fluorobenzaldehyde | |
| 2724908 | |
| ≥98.0% (GC) | |
| 4-Chloro-2-fluorobenzaldehyde |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator