missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluorobenzaldehyde, 99%

Catalog No. AC290620050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC290620050 5 g Glass bottle
AC290620250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC290620050 Supplier Thermo Scientific Chemicals Supplier No. 290620050

CAS: 61072-56-8 | C7H4ClFO | 158.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61072-56-8
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
InChI Key UVGYSEIWAOOIJR-UHFFFAOYSA-N
Synonym 2-fluoro-4-chlorobenzaldehyde, benzaldehyde, 4-chloro-2-fluoro, 4-chloranyl-2-fluoranyl-benzaldehyde, pubchem1435, acmc-1b5ai, ksc494c2f, 3-fluoro-4-formylchlorobenzene, 4-chloro-2-fluoro-benzaldehyde, 4-chloro-2-fluoro-benzoaldehyde, timtec-bb sbb003985
PubChem CID 2724908
IUPAC Name 4-chloro-2-fluorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)F)C=O

Specifications

Melting Point 58.0°C to 61.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H3(F)CHO
Quantity 5 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in most organic solvents
Formula Weight 158.56
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material 4-Chloro-2-fluorobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only