missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-dimethoxyaniline, 98%

CAS: 6358-64-1 | C8H10ClNO2 | 187.62 g/mol

Supplier:  Thermo Scientific Chemicals L0420222

 View more versions of this product

Catalog No. AAL0420222


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chloro-2,5-dimethoxyaniline
118°C to 121°C
C8H10ClNO2
100 g
2,5-dimethoxy-4-chloroaniline, benzenamine, 4-chloro-2,5-dimethoxy, aniline, 4-chloro-2,5-dimethoxy, unii-t659iw8p4s, 4-chloro-2,5-dimethoxyphenylamine, pubchem4366, acmc-1b2t2, dsstox_cid_20718, dsstox_rid_79565, dsstox_gsid_40718
COC1=CC(Cl)=C(OC)C=C1N
187.62
187.63
6358-64-1
163°C (325°F)
MFCD00014893
880445
YGUFQYGSBVXPMC-UHFFFAOYSA-N
4-chloro-2,5-dimethoxyaniline
22833
98%

Chemical Identifiers

6358-64-1
187.62
YGUFQYGSBVXPMC-UHFFFAOYSA-N
22833
COC1=CC(Cl)=C(OC)C=C1N
C8H10ClNO2
MFCD00014893
2,5-dimethoxy-4-chloroaniline, benzenamine, 4-chloro-2,5-dimethoxy, aniline, 4-chloro-2,5-dimethoxy, unii-t659iw8p4s, 4-chloro-2,5-dimethoxyphenylamine, pubchem4366, acmc-1b2t2, dsstox_cid_20718, dsstox_rid_79565, dsstox_gsid_40718
4-chloro-2,5-dimethoxyaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H373
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 228-782-0

RTECSNumber : BX1225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only