missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-dimethoxyaniline, 98%

Catalog No. p-7044040
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0420214 25 g
AAL0420222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0420214 Supplier Thermo Scientific Chemicals Supplier No. L0420214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6358-64-1 | C8H10ClNO2 | 187.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6358-64-1
Molecular Formula C8H10ClNO2
Molecular Weight (g/mol) 187.62
MDL Number MFCD00014893
InChI Key YGUFQYGSBVXPMC-UHFFFAOYSA-N
Synonym 2,5-dimethoxy-4-chloroaniline, benzenamine, 4-chloro-2,5-dimethoxy, aniline, 4-chloro-2,5-dimethoxy, unii-t659iw8p4s, 4-chloro-2,5-dimethoxyphenylamine, pubchem4366, acmc-1b2t2, dsstox_cid_20718, dsstox_rid_79565, dsstox_gsid_40718
PubChem CID 22833
IUPAC Name 4-chloro-2,5-dimethoxyaniline
SMILES COC1=CC(Cl)=C(OC)C=C1N

Specifications

Melting Point 118°C to 121°C
Flash Point 163°C (325°F)
Quantity 25 g
Beilstein 880445
Formula Weight 187.63
Percent Purity 98%
Chemical Name or Material 4-Chloro-2,5-dimethoxyaniline
Hazard Category H302
Hazard Statement GHS H Statement
H301-H373
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 228-782-0
RTECSNumber BX1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only