Learn More
4-Chloro-2,5-dimethoxyaniline, 98%
CAS: 6358-64-1 | C8H10ClNO2 | 187.62 g/mol
Supplier: Thermo Scientific Chemicals L0420214
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 4-Chloro-2,5-dimethoxyaniline | |
| 118°C to 121°C | |
| C8H10ClNO2 | |
| 25 g | |
| 2,5-dimethoxy-4-chloroaniline, benzenamine, 4-chloro-2,5-dimethoxy, aniline, 4-chloro-2,5-dimethoxy, unii-t659iw8p4s, 4-chloro-2,5-dimethoxyphenylamine, pubchem4366, acmc-1b2t2, dsstox_cid_20718, dsstox_rid_79565, dsstox_gsid_40718 | |
| COC1=CC(Cl)=C(OC)C=C1N | |
| 187.62 | |
| 187.63 |
| 6358-64-1 | |
| 163°C (325°F) | |
| MFCD00014893 | |
| 880445 | |
| YGUFQYGSBVXPMC-UHFFFAOYSA-N | |
| 4-chloro-2,5-dimethoxyaniline | |
| 22833 | |
| 98% |
Chemical Identifiers
| 6358-64-1 | |
| 187.62 | |
| YGUFQYGSBVXPMC-UHFFFAOYSA-N | |
| 22833 | |
| COC1=CC(Cl)=C(OC)C=C1N |
| C8H10ClNO2 | |
| MFCD00014893 | |
| 2,5-dimethoxy-4-chloroaniline, benzenamine, 4-chloro-2,5-dimethoxy, aniline, 4-chloro-2,5-dimethoxy, unii-t659iw8p4s, 4-chloro-2,5-dimethoxyphenylamine, pubchem4366, acmc-1b2t2, dsstox_cid_20718, dsstox_rid_79565, dsstox_gsid_40718 | |
| 4-chloro-2,5-dimethoxyaniline |
Safety and Handling
GHS H Statement
H301-H373
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 228-782-0
RTECSNumber : BX1225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only