missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2,5-difluorobenzoic acid, 97%, Thermo Scientific™

Catalog No. p-86795
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC309690100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC309690100 Supplier Thermo Scientific Chemicals Supplier No. 309690100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132794-07-1
Molecular Formula C7H2ClF2O2
Molecular Weight (g/mol) 191.54
MDL Number MFCD00077480
InChI Key PEPCYJSDHYMIFN-UHFFFAOYSA-M
Synonym 2,5-difluoro-4-chlorobenzoic acid, 4-chloro-2,5-difluorobenzoicacid, 4-chloro-2,5-difluoro-benzoic acid, benzoic acid, 4-chloro-2,5-difluoro, pubchem1367, intermediates-zcf02018, acmc-1c2s0, 2,5-difluoro-4-chlorobenzoicacid, attercop-chm at136451, 2,5-difluoro-4-chloro-benzoic acid
PubChem CID 2733274
IUPAC Name 4-chloro-2,5-difluorobenzoic acid
SMILES [O-]C(=O)C1=CC(F)=C(Cl)C=C1F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 153°C to 158°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClC6H2F2(COOH)
Quantity 10 g
Formula Weight 192.55
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-2, 5-difluorobenzoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only