accessibility menu, dialog, popup

UserName

CAS RN 132794-07-1

IUPAC Name: 4-chloro-2,5-difluorobenzoate
Synonyms: 4-chloro-2,5-difluorobenzoate
Molecular Weight (g/mol): 191.54
Molecular Formula: C7H2ClF2O2
InChi Key: PEPCYJSDHYMIFN-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results