missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxy-3-fluorobenzeneboronic acid, 98%

Catalog No. p-7035567
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5328503 1 g
AAH5328506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5328503 Supplier Thermo Scientific Chemicals Supplier No. H5328503
Only null left

CAS: 120153-08-4 | C7H6BFO4 | 183.929 g/mol

4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 120153-08-4
Molecular Formula C7H6BFO4
Molecular Weight (g/mol) 183.929
MDL Number MFCD01114671
InChI Key CZDWJVSOQOMYGC-UHFFFAOYSA-N
Synonym 4-carboxy-3-fluorophenylboronic acid, 4-carboxy-3-fluorobenzeneboronic acid, 4-dihydroxyboranyl-2-fluorobenzoic acid, 4-carboxy-3-fluorophenyl boronic acid, benzoic acid, 4-borono-2-fluoro, 3-fluoro-4-carboxyphenylboronic acid, pubchem5148, acmc-209a6f, benzoic acid,4-borono-2-fluoro, 3-fluoro-4-carboxy-phenylboronic acid
PubChem CID 2763201
IUPAC Name 4-borono-2-fluorobenzoic acid
SMILES B(C1=CC(=C(C=C1)C(=O)O)F)(O)O

Specifications

Melting Point 236°C to 240°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 183.93
Percent Purity 98%
Chemical Name or Material 4-Carboxy-3-fluorobenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only