accessibility menu, dialog, popup

UserName

HOBOHOHOF

CAS RN 120153-08-4

IUPAC Name: 4-(dihydroxyboranyl)-2-fluorobenzoic acid
Synonyms: 4-(dihydroxyboranyl)-2-fluorobenzoic acid
Molecular Weight (g/mol): 183.93
Molecular Formula: C7H6BFO4
InChi Key: CZDWJVSOQOMYGC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C(O)=O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results