missing translation for 'onlineSavingsMsg'
Learn More

4-Carboxy-2-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog No. p-7054111
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62028MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH62028MD Supplier Thermo Scientific Chemicals Supplier No. H62028MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 1050423-87-4
Molecular Formula C13H16BFO4
Molecular Weight (g/mol) 266.075
MDL Number MFCD12756466
InChI Key JSQNZNXVVRPGTD-UHFFFAOYSA-N
Synonym 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxy-2-fluorophenylboronic acid pinacol ester, 4-carboxy-2-fluorobenzeneboronic acid pinacol ester, 4-carboxy-2-fluorophenylboronic acid, pinacol ester, 3-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 4-carboxy-2-fluorophenylboronicacidpinacolester, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoicacid, 3-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl benzoic acid
PubChem CID 46739808
IUPAC Name 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=C(C=C2)C(=O)O)F

Specifications

Melting Point 199°C to 204°C
Quantity 250 mg
Formula Weight 266.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Carboxy-2-fluorobenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures