accessibility menu, dialog, popup

UserName

CAS RN 1050423-87-4

IUPAC Name: 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Synonyms: 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular Weight (g/mol): 266.08
Molecular Formula: C13H16BFO4
InChi Key: JSQNZNXVVRPGTD-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results