missing translation for 'onlineSavingsMsg'
Learn More

4-Butoxybenzoic Acid 98.0+%, TCI America™

Catalog No. B089625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B089625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B089625G Supplier TCI America Supplier No. B089625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1498-96-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00002546
InChI Key LAUFPZPAKULAGB-UHFFFAOYSA-N
Synonym p-butoxybenzoic acid, 4-n-butoxybenzoic acid, benzoic acid, 4-butoxy, benzoic acid, p-butoxy, p-butyloxybenzoic acid, 4-butoxy-benzoic acid, 4-butyloxybenzoic acid, p-n-butoxybenzoic acid, p-n-butyloxybenzoic acid, 4-n-butyloxybenzoic acid
PubChem CID 72971
IUPAC Name 4-butoxybenzoic acid
SMILES CCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 162°C
Color White
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Butoxybenzoic Acid