missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiophenol, 98%

Catalog No. AAA1055509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1055509 10 g
AAA1055518 50 g
AAA1055530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1055509 Supplier Thermo Scientific Chemicals Supplier No. A1055509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 106-53-6 | C6H5BrS | 189.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-53-6
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00004845
InChI Key FTBCOQFMQSTCQQ-UHFFFAOYSA-N
Synonym 4-bromothiophenol, benzenethiol, 4-bromo, p-bromothiophenol, p-bromobenzenethiol, 4-bromobenzene-1-thiol, 4-bromo-benzenethiol, 4-bromo thiophenol, p-bromophenyl thiol, benzenethiol, p-bromo, unii-p3wuo03x17
PubChem CID 66049
IUPAC Name 4-bromobenzenethiol
SMILES C1=CC(=CC=C1S)Br

Specifications

Melting Point 72°C to 78°C
Boiling Point 239°C
Odor Unpleasant
Quantity 10 g
UN Number UN2923
Beilstein 507031
Sensitivity Air Sensitive
Formula Weight 189.08
Percent Purity 98%
Chemical Name or Material 4-Bromothiophenol
Hazard Category H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-407-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only