accessibility menu, dialog, popup

UserName

CAS RN 106-53-6

IUPAC Name: 4-bromobenzene-1-thiol
Synonyms: benzenethiol, 4-bromo-
Molecular Weight (g/mol): 189.07
Molecular Formula: C6H5BrS
InChi Key: FTBCOQFMQSTCQQ-UHFFFAOYSA-N
SMILES: SC1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)