missing translation for 'onlineSavingsMsg'
Learn More

4-Bromothiophenol, 95%

Catalog No. p-77367
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC107460250 25 g Glass bottle
AC107461000 100 g Glass bottle
2 options

Catalog No. AC107460250

Supplier: Thermo Scientific Chemicals 107460250

Only null left

CAS: 106-53-6 | C6H5BrS | 189.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 106-53-6
Molecular Formula C6H5BrS
Molecular Weight (g/mol) 189.07
MDL Number MFCD00004845
InChI Key FTBCOQFMQSTCQQ-UHFFFAOYSA-N
Synonym 4-bromothiophenol, benzenethiol, 4-bromo, p-bromothiophenol, p-bromobenzenethiol, 4-bromobenzene-1-thiol, 4-bromo-benzenethiol, 4-bromo thiophenol, p-bromophenyl thiol, benzenethiol, p-bromo, unii-p3wuo03x17
PubChem CID 66049
IUPAC Name 4-bromobenzenethiol
SMILES C1=CC(=CC=C1S)Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 71°C to 76°C
Color White to Beige
Boiling Point 239°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula BrC6H4SH
Quantity 25 g
Beilstein 06, 330
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alkaline solutions (reacts),soluble in methanol
Formula Weight 189.07
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Bromothiophenol, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Toxic in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if inhaled.
Toxic if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-407-3
TSCA TSCA

RUO – Research Use Only