missing translation for 'onlineSavingsMsg'
Learn More

4-Bromotetraphenylsilane 96.0+%, TCI America™

Catalog No. B3287200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3287200MG 200 mg
B32871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3287200MG Supplier TCI America Supplier No. B3287200MG
Only null left

Chemical Identifiers

CAS 18737-40-1
Molecular Formula C24H19BrSi
Molecular Weight (g/mol) 415.405
InChI Key UDZSLJULKCKKPX-UHFFFAOYSA-N
PubChem CID 297506
IUPAC Name (4-bromophenyl)-triphenylsilane
SMILES C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)Br

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 200 mg
Formula Weight 415.41
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromotetraphenylsilane