CAS RN 18737-40-1
IUPAC Name:
(4-bromophenyl)triphenylsilane
Synonyms:
(4-bromophenyl)triphenylsilane
Molecular Weight (g/mol):
415.41
Molecular Formula:
C24H19BrSi
InChi Key:
UDZSLJULKCKKPX-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4-Bromotetraphenylsilane 96.0+%, TCI America™
CAS: 18737-40-1 Molecular Formula: C24H19BrSi Molecular Weight (g/mol): 415.405 InChI Key: UDZSLJULKCKKPX-UHFFFAOYSA-N PubChem CID: 297506 IUPAC Name: (4-bromophenyl)-triphenylsilane SMILES: C1=CC=C(C=C1)[Si](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=C(C=C4)Br