missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophthalonitrile 97.0+%, TCI America™

Catalog No. p-7124103
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B55091G 1 g
B55095G 5 g
2 options

Catalog No. B55091G

Supplier: TCI America B55091G

Only null left

Chemical Identifiers

CAS 70484-01-4
Molecular Formula C8H3BrN2
Molecular Weight (g/mol) 207.03
MDL Number MFCD00221794
InChI Key ARAONWRSVQOHIT-UHFFFAOYSA-N
Synonym 4-Bromo-1,2-dicyanobenzene
PubChem CID 11356223
IUPAC Name 4-bromobenzene-1,2-dicarbonitrile
SMILES BrC1=CC(C#N)=C(C=C1)C#N

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 1 g
UN Number 3439
Formula Weight 207.03
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophthalonitrile