accessibility menu, dialog, popup

UserName

CAS RN 70484-01-4

BrNN

CAS RN 70484-01-4

IUPAC Name: 4-bromobenzene-1,2-dicarbonitrile
Synonyms: 4-bromobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 207.03
Molecular Formula: C8H3BrN2
InChi Key: ARAONWRSVQOHIT-UHFFFAOYSA-N
SMILES: BrC1=CC(C#N)=C(C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results