missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AAH58429AD
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58429AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58429AD Supplier Thermo Scientific Chemicals Supplier No. H58429AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 148651-39-2 | C6H4BrIZn | 348.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148651-39-2
Molecular Formula C6H4BrIZn
Molecular Weight (g/mol) 348.286
MDL Number MFCD01311441
InChI Key PLIIAGJFCAKUCV-UHFFFAOYSA-M
Synonym 4-bromophenylzinc iodide, 4-bromophenylzinc iodide solution, 4-bromophenyl iodo zinc, 4-bromophenylzinc iodide 0.5 m in tetrahydrofuran, 4-bromophenylzinc iodide solution, 0.5 m in thf, 4-bromophenylzinc iodide, 0.5m in thf, packaged under argon in resealable chemseal bottles
PubChem CID 4595200
IUPAC Name bromobenzene;iodozinc(1+)
SMILES C1=CC(=CC=[C-]1)Br.[Zn+]I

Specifications

Density 1.045
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Formula Weight 348.29
Chemical Name or Material 4-Bromophenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Hazard Category H225-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H261-H314-H351-H335
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only