CAS RN 148651-39-2
IUPAC Name:
4-bromobenzen-1-ide; iodozincylium
Synonyms:
4-bromobenzen-1-ide; iodozincylium
Molecular Weight (g/mol):
348.29
Molecular Formula:
C6H4BrIZn
InChi Key:
PLIIAGJFCAKUCV-UHFFFAOYSA-M
SMILES:
[Zn+]I.BrC1=CC=[C-]C=C1
1
–
1
of
1
results
1
4-Bromophenylzinc iodide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
CAS: 148651-39-2 Molecular Formula: C6H4BrIZn Molecular Weight (g/mol): 348.286 MDL Number: MFCD01311441 InChI Key: PLIIAGJFCAKUCV-UHFFFAOYSA-M Synonym: 4-bromophenylzinc iodide,4-bromophenylzinc iodide solution,4-bromophenyl iodo zinc,4-bromophenylzinc iodide 0.5 m in tetrahydrofuran,4-bromophenylzinc iodide solution, 0.5 m in thf,4-bromophenylzinc iodide, 0.5m in thf, packaged under argon in resealable chemseal bottles PubChem CID: 4595200 IUPAC Name: bromobenzene;iodozinc(1+) SMILES: C1=CC(=CC=[C-]1)Br.[Zn+]I