missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromophenylacetone 98.0+%, TCI America™
Supplier: TCI America B438725G
Specifications
| 4-Bromophenylacetone | |
| Yellow | |
| C24H24N2O5S | |
| 25 g | |
| CDUPPXGKUKTGLU-UHFFFAOYSA-N | |
| methyl 2-{2-[N-(3,5-dimethylphenyl)benzenesulfonamido]acetamido}benzoate | |
| 736332 | |
| ≥98.0% (GC) |
| 6186-22-7 | |
| 139°C | |
| MFCD00210401 | |
| 4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone | |
| COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1 | |
| 452.53 | |
| 213.07 | |
| Liquid |
Chemical Identifiers
| 6186-22-7 | |
| 452.53 | |
| CDUPPXGKUKTGLU-UHFFFAOYSA-N | |
| 736332 | |
| COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1 |
| C24H24N2O5S | |
| MFCD00210401 | |
| 4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone | |
| methyl 2-{2-[N-(3,5-dimethylphenyl)benzenesulfonamido]acetamido}benzoate |
Safety and Handling
TSCA : No