missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenylacetone 98.0+%, TCI America™

Catalog No. B43875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43875G 5 g
B438725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43875G Supplier TCI America Supplier No. B43875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6186-22-7
Molecular Formula C24H24N2O5S
Molecular Weight (g/mol) 452.53
MDL Number MFCD00210401
InChI Key CDUPPXGKUKTGLU-UHFFFAOYSA-N
Synonym 4-bromophenylacetone, 1-4-bromophenyl propan-2-one, 1-4-bromophenyl acetone, 1-4-bromophenyl-2-propanone, 1-4-bromo-phenyl-propan-2-one, p-bromo benzyl methyl ketone, 4-bromophenylaceton, 4-bromophenylacteone, 4-bromophenyl acetone
PubChem CID 736332
IUPAC Name methyl 2-{2-[N-(3,5-dimethylphenyl)benzenesulfonamido]acetamido}benzoate
SMILES COC(=O)C1=CC=CC=C1NC(=O)CN(C1=CC(C)=CC(C)=C1)S(=O)(=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 139°C
Quantity 5 g
Formula Weight 213.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromophenylacetone