missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromophenyl Methyl Sulfone 98.0+%, TCI America™
Supplier: TCI America B314825G
Specifications
| 4-Bromophenyl Methyl Sulfone | |
| 104°C | |
| C7H7BrO2S | |
| 25 g | |
| FJLFSYRGFJDJMQ-UHFFFAOYSA-N | |
| 1-bromo-4-methylsulfonylbenzene | |
| 77014 | |
| ≥98.0% (GC) |
| 3466-32-8 | |
| White-Yellow | |
| MFCD00025065 | |
| 4-bromophenyl methyl sulfone, 1-bromo-4-methylsulfonyl benzene, 1-bromo-4-methanesulfonylbenzene, p-bromophenyl methyl sulfone, methyl 4-bromophenyl sulfone, 4-bromophenyl methyl sulphone, benzene, 1-bromo-4-methylsulfonyl, 4-bromophenylmethylsulfone, p-methyl benzenesulfobromide, sulfone, p-bromophenyl methyl | |
| CS(=O)(=O)C1=CC=C(C=C1)Br | |
| 235.095 | |
| 235.10 | |
| Crystalline Powder |
Chemical Identifiers
| 3466-32-8 | |
| 235.095 | |
| FJLFSYRGFJDJMQ-UHFFFAOYSA-N | |
| 77014 | |
| CS(=O)(=O)C1=CC=C(C=C1)Br |
| C7H7BrO2S | |
| MFCD00025065 | |
| 4-bromophenyl methyl sulfone, 1-bromo-4-methylsulfonyl benzene, 1-bromo-4-methanesulfonylbenzene, p-bromophenyl methyl sulfone, methyl 4-bromophenyl sulfone, 4-bromophenyl methyl sulphone, benzene, 1-bromo-4-methylsulfonyl, 4-bromophenylmethylsulfone, p-methyl benzenesulfobromide, sulfone, p-bromophenyl methyl | |
| 1-bromo-4-methylsulfonylbenzene |
Safety and Handling
TSCA : No