missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl Methyl Sulfone 98.0+%, TCI America™

Catalog No. p-7119984
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31485G 5 g
B314825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31485G Supplier TCI America Supplier No. B31485G
Only null left

Chemical Identifiers

CAS 3466-32-8
Molecular Formula C7H7BrO2S
Molecular Weight (g/mol) 235.095
MDL Number MFCD00025065
InChI Key FJLFSYRGFJDJMQ-UHFFFAOYSA-N
Synonym 4-bromophenyl methyl sulfone, 1-bromo-4-methylsulfonyl benzene, 1-bromo-4-methanesulfonylbenzene, p-bromophenyl methyl sulfone, methyl 4-bromophenyl sulfone, 4-bromophenyl methyl sulphone, benzene, 1-bromo-4-methylsulfonyl, 4-bromophenylmethylsulfone, p-methyl benzenesulfobromide, sulfone, p-bromophenyl methyl
PubChem CID 77014
IUPAC Name 1-bromo-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)Br

Specifications

Melting Point 104°C
Color White-Yellow
Quantity 5 g
Formula Weight 235.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenyl Methyl Sulfone