missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromophenyl Isothiocyanate 98.0+%, TCI America™
Supplier: TCI America I052625G
Specifications
| 4-Bromophenyl Isothiocyanate | |
| 58°C | |
| C7H4BrNS | |
| 25 g | |
| 4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate | |
| C1=CC(=CC=C1N=C=S)Br | |
| 214.08 | |
| 214.08 | |
| Crystalline Powder |
| 1985-12-2 | |
| White-Yellow | |
| MFCD00004808 | |
| 1759 | |
| XQACWEBGSZBLRG-UHFFFAOYSA-N | |
| 1-bromo-4-isothiocyanatobenzene | |
| 16133 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 1985-12-2 | |
| 214.08 | |
| XQACWEBGSZBLRG-UHFFFAOYSA-N | |
| 16133 | |
| C1=CC(=CC=C1N=C=S)Br |
| C7H4BrNS | |
| MFCD00004808 | |
| 4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate | |
| 1-bromo-4-isothiocyanatobenzene |
Safety and Handling
RTECSNumber : NX8400000
TSCA : No