Learn More
4-Bromophenyl isothiocyanate, 97%
CAS: 1985-12-2 | C7H4BrNS | 214.08 g/mol
$107.52 - $107.52
Chemical Identifiers
| CAS | 1985-12-2 |
|---|---|
| Molecular Formula | C7H4BrNS |
| Molecular Weight (g/mol) | 214.08 |
| MDL Number | MFCD00004808 |
| InChI Key | XQACWEBGSZBLRG-UHFFFAOYSA-N |
| Synonym | 4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate |
| PubChem CID | 16133 |
| IUPAC Name | 1-bromo-4-isothiocyanatobenzene |
| SMILES | C1=CC(=CC=C1N=C=S)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1110106
|
Thermo Scientific Chemicals
L1110106 |
5 g |
Each for $107.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1985-12-2 | |
| 214.08 | |
| XQACWEBGSZBLRG-UHFFFAOYSA-N | |
| 16133 | |
| C1=CC(=CC=C1N=C=S)Br |
| C7H4BrNS | |
| MFCD00004808 | |
| 4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate | |
| 1-bromo-4-isothiocyanatobenzene |
Specifications
| 1985-12-2 | |
| 144°C to 145°C (5 mmHg) | |
| C7H4BrNS | |
| 5 g | |
| 878549 | |
| 4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate | |
| C1=CC(=CC=C1N=C=S)Br | |
| 214.08 | |
| 214.09 | |
| 4-Bromophenyl isothiocyanate |
| 56°C to 60°C | |
| >110°C (230°F) | |
| MFCD00004808 | |
| UN1759 | |
| Moisture sensitive | |
| XQACWEBGSZBLRG-UHFFFAOYSA-N | |
| 1-bromo-4-isothiocyanatobenzene | |
| 16133 | |
| 97% |
Safety and Handling
GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 000-000-0
RTECSNumber : NX8400000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only