missing translation for 'onlineSavingsMsg'
Learn More

4-Bromophenyl isothiocyanate, 97%

CAS: 1985-12-2 | C7H4BrNS | 214.08 g/mol

Supplier:  Thermo Scientific Chemicals L1110106

 View more versions of this product

Catalog No. AAL1110106


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromophenyl isothiocyanate
56°C to 60°C
>110°C (230°F)
MFCD00004808
UN1759
Moisture sensitive
XQACWEBGSZBLRG-UHFFFAOYSA-N
1-bromo-4-isothiocyanatobenzene
16133
97%
1985-12-2
144°C to 145°C (5 mmHg)
C7H4BrNS
5 g
878549
4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
C1=CC(=CC=C1N=C=S)Br
214.08
214.09

Chemical Identifiers

1985-12-2
214.08
XQACWEBGSZBLRG-UHFFFAOYSA-N
16133
C1=CC(=CC=C1N=C=S)Br
C7H4BrNS
MFCD00004808
4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
1-bromo-4-isothiocyanatobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

EINECSNumber : 000-000-0

RTECSNumber : NX8400000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only