missing translation for 'onlineSavingsMsg'
Learn More

4-(Bromomethyl)benzophenone 96.0+%, TCI America™

Catalog No. B38405G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B38405G Supplier TCI America Supplier No. B38405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 32752-54-8
Molecular Formula C14H11BrO
Molecular Weight (g/mol) 275.145
MDL Number MFCD00092076
InChI Key RYULULVJWLRDQH-UHFFFAOYSA-N
PubChem CID 122951
IUPAC Name [4-(bromomethyl)phenyl]-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)CBr

Specifications

Melting Point 112°C
Color Yellow
Quantity 5 g
Formula Weight 275.15
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Bromomethyl)benzophenone